siRNA Details
Gene Symbol: Gzmg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010375.2 | NP_034505.1 | 2 | 196 | ||
GenBank mRNA | BC099473.1 | - | 2 | 196 | 30715974 | |
J02872.1 | - | 2 | 182 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gzmg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010375.2 | NP_034505.1 | 2 | 196 | ||
GenBank mRNA | BC099473.1 | - | 2 | 196 | 30715974 | |
J02872.1 | - | 2 | 182 |