siRNA Details
| Gene Symbol: H2-Ob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010389.3 | NP_034519.2 | 2 | 189 | ||
| GenBank mRNA | AK053233.1 | - | 2 | 238 | ||
| AK155144.1 | - | 2 | 139 | |||
| AK155650.1 | - | 2 | 189 | |||
| AK172191.1 | - | 2 | 194 | |||
| AK172273.1 | - | 2 | 164 | |||
| AK172299.1 | - | 2 | 164 | |||
| BC127027.1 | - | 2 | 122 | 40044586 | ||
| BC127028.1 | - | 2 | 122 | 40044587 | ||
| BY205160.1 | - | 2 | 190 | |||
| KJ650230.1 | - | 2 | 122 | |||
| KJ663722.1 | - | 2 | 122 | |||

H2-Ob
Genomic Map