siRNA Details
| Gene Symbol: Gp1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010326.2 | NP_034456.2 | 2 | 681 | ||
| GenBank mRNA | BC137765.1 | - | 2 | 661 | 8860784 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gp1ba| Gene Symbol: Gp1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010326.2 | NP_034456.2 | 2 | 681 | ||
| GenBank mRNA | BC137765.1 | - | 2 | 661 | 8860784 | |