siRNA Details
| Gene Symbol: Gzmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010372.2 | NP_034502.2 | 2 | 233 | ||
| GenBank mRNA | AK131671.1 | - | 2 | 234 | ||
| BC141122.1 | - | 2 | 188 | 9055734 | ||
| BC145433.1 | - | 188 | 9053991 | |||
| BC145434.1 | - | 2 | 188 | |||
| J03255.1 | - | 2 | 248 | |||

Gzmd
Genomic Map