siRNA Details
| Gene Symbol: H2-Ob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010389.3 | NP_034519.2 | 2 | 258 | ||
| GenBank mRNA | AK053233.1 | - | 2 | 307 | ||
| AK155144.1 | - | 2 | 208 | |||
| AK155650.1 | - | 2 | 258 | |||
| AK172191.1 | - | 2 | 263 | |||
| AK172273.1 | - | 2 | 233 | |||
| AK172299.1 | - | 2 | 233 | |||
| BC100707.1 | - | 2 | 187 | |||
| BC100709.1 | - | 2 | 184 | |||
| BC100710.1 | - | 2 | 186 | |||
| BC127027.1 | - | 2 | 191 | 40044586 | ||
| BC127028.1 | - | 2 | 191 | 40044587 | ||
| BY205160.1 | - | 2 | 259 | |||
| KJ650230.1 | - | 2 | 191 | |||
| KJ663722.1 | - | 2 | 191 | |||

H2-Ob
Genomic Map