siRNA Details
| Gene Symbol: H2-T10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010395.7 | NP_034525.4 | 974 | |||
| GenBank mRNA | AK088241.1 | - | 3 | 476 | ||
| AK153876.1 | - | 963 | ||||
| AK161731.1 | - | 3 | 1290 | |||
| BC021751.1 | - | 1068 | 4165944 | |||
| BC021760.1 | - | 1092 | 5055493 | |||
| M35244.1 | - | 4 | 1078 | |||
| M35246.1 | - | 5 | 966 | |||

H2-T10
Genomic Map