siRNA Details
| Gene Symbol: Gzmc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010371.2 | NP_034501.2 | 4 | 592 | ||
| GenBank mRNA | BC120755.1 | - | 4 | 542 | 40129678 | |
| BC120757.1 | - | 4 | 542 | 40129680 | ||
| BC145030.1 | - | 4 | 542 | 9053578 | ||
| M12301.1 | - | 1 | 46 | |||
| M18459.1 | - | 4 | 592 | |||
| X12822.1 | - | 4 | 536 | |||

Gzmc
Genomic Map