siRNA Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010431.2 | NP_034561.2 | 2 | 613 | ||
| GenBank mRNA | AF003695.1 | - | 2 | 447 | ||
| AK017853.1 | - | 2 | 474 | |||
| AK034087.1 | - | 2 | 579 | |||
| AK138944.1 | - | 2 | 480 | |||
| AK150367.1 | - | 2 | 480 | |||
| BC026139.1 | - | 2 | 465 | 4019056 | ||
| BY725437.1 | - | 2 | 613 | |||
| X95002.1 | - | 1 | 127 | |||
| X95580.1 | - | 2 | 279 | |||

Hif1a
Genomic Map