siRNA Details
| Gene Symbol: Hipk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145824.1 | NP_001139296.1 | 2 | 660 | ||
| NM_010434.2 | NP_034564.2 | 2 | 660 | |||
| GenBank mRNA | AF077660.1 | - | 2 | 414 | ||
| AF170305.1 | - | 1 | 124 | |||
| AF305238.1 | - | 1 | 122 | |||
| AK140130.1 | - | 2 | 763 | |||
| BC131663.1 | - | 2 | 291 | |||
| BC131664.1 | - | 2 | 291 | 40110751 | ||

Hipk3
Genomic Map