siRNA Details
| Gene Symbol: Hoxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010451.2 | NP_034581.1 | 2 | 798 | ||
| GenBank mRNA | AB221621.1 | - | 2 | 615 | ||
| AK134501.1 | - | 2 | 447 | |||
| AK157713.1 | - | 2 | 413 | |||
| BC117105.1 | - | 2 | 745 | 40126424 | ||
| BC117107.1 | - | 2 | 745 | 40126426 | ||
| M93292.1 | - | 2 | 709 | |||
| M95599.1 | - | 2 | 798 | |||

Hoxa2
Genomic Map