siRNA Details
| Gene Symbol: Hoxa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010454.1 | NP_034584.1 | 1 | 145 | ||
| GenBank mRNA | AB221546.1 | - | 1 | 145 | ||
| BC119105.1 | - | 1 | 243 | 8733854 | ||
| BC119107.1 | - | 1 | 243 | 8733856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxa6| Gene Symbol: Hoxa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010454.1 | NP_034584.1 | 1 | 145 | ||
| GenBank mRNA | AB221546.1 | - | 1 | 145 | ||
| BC119105.1 | - | 1 | 243 | 8733854 | ||
| BC119107.1 | - | 1 | 243 | 8733856 | ||