siRNA Details
| Gene Symbol: Hoxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010469.2 | NP_034599.2 | 5, 6 | 1653 | ||
| GenBank mRNA | AK135479.1 | - | 1, 2 | 444 | ||
| BC139206.1 | - | 1, 2 | 545 | 8862228 | ||
| BC139207.1 | - | 1, 2 | 545 | 8862229 | ||

Hoxd4
Genomic Map