siRNA Details
| Gene Symbol: Htr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010482.1 | NP_034612.1 | 1 | 578 | ||
| GenBank mRNA | BC119129.1 | - | 1 | 641 | 8733878 | |
| BC131951.1 | - | 1 | 641 | 40130228 | ||
More Information
Set Membership: |
|

Htr1b
Genomic Map