siRNA Details
| Gene Symbol: Ica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252266.1 | NP_001239195.1 | 2 | 186 | ||
| NM_010492.3 | NP_034622.3 | 3 | 599 | |||
| GenBank mRNA | AB303567.1 | - | 2 | 110 | ||
| AK005965.1 | - | 3 | 304 | |||
| AK016989.1 | - | 3 | 302 | |||
| BC002030.1 | - | 3 | 233 | 3487814 | ||
| CF914747.1 | - | 3 | 302 | |||
| U26459.1 | - | 3 | 222 | |||
| U26460.1 | - | 2 | 128 | |||
| U26461.1 | - | 2 | 127 | |||
| U34595.1 | - | 1 | 95 | |||

Ica1
Genomic Map