siRNA Details
| Gene Symbol: Ifnar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110498.1 | NP_001103968.1 | 5 | 581 | ||
| NM_010509.2 | NP_034639.2 | 5 | 581 | |||
| GenBank mRNA | AF013274.1 | - | 5 | 359 | ||
| AF013486.1 | - | 5 | 453 | |||
| AK003185.1 | - | 5 | 515 | |||
| AK010952.1 | - | 5 | 532 | |||
| AK139169.1 | - | 5 | 561 | |||
| AK150386.1 | - | 5 | 515 | |||
| AK150597.1 | - | 5 | 494 | |||
| AK151474.1 | - | 5 | 509 | |||
| AK151493.1 | - | 5 | 494 | |||
| AK151777.1 | - | 5 | 509 | |||
| AK152878.1 | - | 5 | 515 | |||
| AK153317.1 | - | 5 | 515 | |||
| AK209089.1 | - | 2 | 61 | |||
| BC003743.1 | - | 4 | 276 | |||
| BC071225.1 | - | 4 | 279 | 4187603 | ||
| Y09813.1 | - | 5 | 336 | |||
| Y09864.1 | - | 5 | 471 | |||
| Y09865.1 | - | 5 | 471 | |||

Ifnar2
Genomic Map