siRNA Details
| Gene Symbol: Ihh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010544.2 | NP_034674.1 | 2 | 769 | ||
| GenBank mRNA | AK090147.1 | - | 2 | 564 | ||
| AK144322.1 | - | 2 | 792 | |||
| BC046984.1 | - | 2 | 769 | 6477118 | ||
| U85610.1 | - | 2 | 629 | |||
| X76291.1 | - | 2 | 107 | |||

Ihh
Genomic Map