siRNA Details
| Gene Symbol: Hoxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010469.2 | NP_034599.2 | 6 | 1893 | ||
| GenBank mRNA | AK135479.1 | - | 2 | 684 | ||
| AK136751.1 | - | 691 | ||||
| BC139206.1 | - | 2 | 785 | 8862228 | ||
| BC139207.1 | - | 2 | 785 | 8862229 | ||
| J03770.1 | - | 2 | 864 | |||

Hoxd4
Genomic Map