siRNA Details
| Gene Symbol: Hpca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130419.2 | NP_001123891.1 | 2, 3 | 585 | ||
| NM_001286081.1 | NP_001273010.1 | 2, 3 | 498 | |||
| NM_001286083.1 | NP_001273012.1 | 2, 3 | 430 | |||
| NM_010471.4 | NP_034601.1 | 2, 3 | 489 | |||
| GenBank mRNA | AF326551.1 | - | 1, 2 | 612 | ||
| AF326552.1 | - | 1, 2 | 612 | |||
| AK002992.1 | - | 2, 3 | 489 | |||
| BC049607.1 | - | 2, 3 | 454 | 6595363 | ||
| BC058588.1 | - | 1, 2 | 403 | 5360894 | ||
| CX238650.1 | - | 2, 3 | 462 | |||
| CX239660.1 | - | 3, 4 | 584 | |||
| CX240088.1 | - | 2, 3 | 430 | |||
| CX240757.1 | - | 2, 3 | 478 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_017122, XM_006238920, XM_006238921, AY442172, D12573, and others.

Hpca
Genomic Map