siRNA Details
| Gene Symbol: Ids | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010498.3 | NP_034628.2 | 3 | 525 | ||
| GenBank mRNA | AF538870.1 | - | 3 | 524 | ||
| AK085237.1 | - | 4 | 402 | |||
| AK132287.1 | - | 4 | 405 | |||
| AK166178.1 | - | 3 | 403 | |||
| AK166443.1 | - | 4 | 354 | |||
| L07921.1 | - | 3 | 402 | |||

Ids
Genomic Map