siRNA Details
| Gene Symbol: Il1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010555.4 | NP_034685.1 | 3 | 269 | ||
| GenBank mRNA | AK132264.1 | - | 3 | 296 | ||
| CT010182.1 | - | 2 | 177 | |||
| X59769.1 | - | 3 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1r2| Gene Symbol: Il1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010555.4 | NP_034685.1 | 3 | 269 | ||
| GenBank mRNA | AK132264.1 | - | 3 | 296 | ||
| CT010182.1 | - | 2 | 177 | |||
| X59769.1 | - | 3 | 269 | |||