siRNA Details
| Gene Symbol: Hoxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010467.2 | NP_034597.2 | 2 | 1252 | ||
| GenBank mRNA | BC120537.1 | - | 2 | 1151 | 40129460 | |
| BC120539.1 | - | 2 | 1151 | 40129462 | ||
| X60034.1 | - | 2 | 1249 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxd1| Gene Symbol: Hoxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010467.2 | NP_034597.2 | 2 | 1252 | ||
| GenBank mRNA | BC120537.1 | - | 2 | 1151 | 40129460 | |
| BC120539.1 | - | 2 | 1151 | 40129462 | ||
| X60034.1 | - | 2 | 1249 | |||