siRNA Details
| Gene Symbol: Hspd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010477.4 | NP_034607.3 | 11 | 1623 | ||
| GenBank mRNA | AK088844.1 | - | 11 | 1537 | ||
| AK143882.1 | - | 11 | 1528 | |||
| AK146831.1 | - | 11 | 1575 | |||
| AK202454.1 | - | 2 | 151 | |||
| BC016400.1 | - | 11 | 1509 | 3585270 | ||
| BC018545.1 | - | 5 | 731 | |||
| BC106112.1 | - | 11 | 1524 | |||
| BF607107.1 | - | 1 | 53 | |||
| X53584.1 | - | 10 | 1413 | |||
| X55023.1 | - | 11 | 1485 | |||

Hspd1
Genomic Map