siRNA Details
Gene Symbol: Htr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010483.3 | NP_034613.2 | 2 | 1667 | ||
GenBank mRNA | X69867.1 | - | 2 | 1702 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
