siRNA Details
| Gene Symbol: Ids | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010498.3 | NP_034628.2 | 5 | 816 | ||
| GenBank mRNA | AJ627044.1 | - | 3 | 281 | ||
| AK085237.1 | - | 6 | 693 | |||
| AK132287.1 | - | 6 | 696 | |||
| L07921.1 | - | 5 | 693 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ids| Gene Symbol: Ids | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010498.3 | NP_034628.2 | 5 | 816 | ||
| GenBank mRNA | AJ627044.1 | - | 3 | 281 | ||
| AK085237.1 | - | 6 | 693 | |||
| AK132287.1 | - | 6 | 696 | |||
| L07921.1 | - | 5 | 693 | |||