siRNA Details
| Gene Symbol: Ier5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010500.2 | NP_034630.1 | 1 | 1960 | ||
| GenBank mRNA | AF079527.1 | - | 1 | 1961 | ||
| AK146609.1 | - | 1 | 1957 | |||
| AK150461.1 | - | 1 | 1959 | |||
| AW913049.1 | - | 1 | 133 | |||
| BC051986.1 | - | 1 | 1934 | 5707524 | ||

Ier5
Genomic Map