siRNA Details
| Gene Symbol: Igf2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010515.2 | NP_034645.2 | 40, 41 | 6221 | ||
| GenBank mRNA | AK053045.1 | - | 5, 6 | 813 | ||
| AK088699.1 | - | 15, 16 | 2447 | |||
| BC031190.1 | - | 7, 8 | 1227 | |||
| BC043457.1 | - | 3, 4 | 422 | |||
| BC055018.1 | - | 3, 4 | 427 | |||
| BC150817.1 | - | 40, 41 | 6132 | 9087728 | ||
| BC171973.1 | - | 40, 41 | 6132 | 9054259 | ||
| U04710.1 | - | 40, 41 | 6177 | |||
| X60389.1 | - | 7, 8 | 1176 | |||

Igf2r
Genomic Map