siRNA Details
| Gene Symbol: Il17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010552.3 | NP_034682.1 | 3 | 653 | ||
| GenBank mRNA | AK040420.1 | - | 3 | 654 | ||
| U43088.1 | - | 3 | 640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17a| Gene Symbol: Il17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010552.3 | NP_034682.1 | 3 | 653 | ||
| GenBank mRNA | AK040420.1 | - | 3 | 654 | ||
| U43088.1 | - | 3 | 640 | |||