siRNA Details
| Gene Symbol: Il3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010556.4 | NP_034686.2 | 3 | 310 | ||
| GenBank mRNA | A02046.1 | - | 3 | 308 | ||
| AK153634.1 | - | 3 | 311 | |||
| BC125552.1 | - | 3 | 305 | 40130067 | ||
| BC125554.1 | - | 3 | 305 | 40130069 | ||
| DQ788721.1 | - | 3 | 281 | |||
| K01668.1 | - | 3 | 308 | |||
| K01850.1 | - | 3 | 308 | |||

Il3
Genomic Map