siRNA Details
Gene Symbol: Inhbc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010565.3 | NP_034695.1 | 2 | 890 | ||
GenBank mRNA | AK131946.1 | - | 2 | 490 | ||
BC026140.1 | - | 2 | 788 | 4212175 | ||
U95962.1 | - | 2 | 890 | |||
X90819.1 | - | 2 | 873 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
