siRNA Details
| Gene Symbol: Jag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010588.2 | NP_034718.2 | 5 | 761 | ||
| GenBank mRNA | AF038572.1 | - | 5 | 761 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jag2| Gene Symbol: Jag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010588.2 | NP_034718.2 | 5 | 761 | ||
| GenBank mRNA | AF038572.1 | - | 5 | 761 | ||