siRNA Details
| Gene Symbol: Kcnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159850.1 | NP_001153322.1 | 2 | 548 | ||
| NM_001281847.1 | NP_001268776.1 | 2 | 439 | |||
| NM_001281848.1 | NP_001268777.1 | 2 | 398 | |||
| NM_010607.3 | NP_034737.2 | 2 | 644 | |||
| GenBank mRNA | AK138427.1 | - | 2 | 320 | ||
| AK140716.1 | - | 2 | 319 | |||
| AK164197.1 | - | 2 | 398 | |||
| AY736359.1 | - | 2 | 283 | |||
| BC062094.1 | - | 2 | 262 | 6848897 | ||
| BY267214.1 | - | 1 | 83 | |||
| U73488.2 | - | 3 | 721 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_172042, NM_172041, XM_006250434, XM_006250433, AY727922, and others.

Kcnk2
Genomic Map