siRNA Details
| Gene Symbol: Kifc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145831.1 | NP_001139303.1 | 8 | 909 | ||
| NM_001145832.1 | NP_001139304.1 | 4 | 393 | |||
| NM_010631.2 | NP_034761.3 | 6 | 736 | |||
| GenBank mRNA | AF013118.2 | - | 2 | 372 | ||
| AK132895.1 | - | 4 | 393 | |||
| BC070429.1 | - | 6 | 736 | 6810893 | ||
| BI737872.1 | - | 4 | 328 | |||
| D49546.1 | - | 6 | 751 | |||

Kifc3
Genomic Map