siRNA Details
| Gene Symbol: Klf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010638.4 | NP_034768.2 | 1 | 702 | ||
| GenBank mRNA | AK028544.1 | - | 1 | 705 | ||
| BC106117.1 | - | 2 | 529 | |||
| BC138724.1 | - | 1 | 520 | 8861746 | ||
| BC138725.1 | - | 1 | 515 | 8861747 | ||
| CT010185.1 | - | 1 | 173 | |||
| Y14296.1 | - | 1 | 173 | |||

Klf9
Genomic Map