siRNA Details
| Gene Symbol: Klk1b21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010642.2 | NP_034772.1 | 3 | 317 | ||
| GenBank mRNA | AB039276.1 | - | 3 | 320 | ||
| AK144221.1 | - | 3 | 284 | |||
| BC012243.1 | - | 3 | 251 | 4167772 | ||
| BC145383.1 | - | 3 | 273 | 9053941 | ||

Klk1b21
Genomic Map