siRNA Details
| Gene Symbol: Klra12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010646.1 | NP_034776.1 | 4, 5 | 626 | ||
| GenBank mRNA | AF204265.1 | - | 5, 6 | 617 | ||
| AF204266.1 | - | 4, 5 | 578 | |||
| AF204267.1 | - | 5, 6 | 608 | |||
| AF204268.1 | - | 4, 5 | 569 | |||
| AF307947.1 | - | 4, 5 | 630 | |||
| AF307948.1 | - | 4, 5 | 626 | |||

Klra12
Genomic Map