siRNA Details
| Gene Symbol: Klrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098669.1 | NP_001092139.1 | 4 | 534 | ||
| NM_010653.4 | NP_034783.3 | 5 | 569 | |||
| GenBank mRNA | AF106010.1 | - | 5 | 702 | ||
| AF106011.2 | - | 4 | 534 | |||
| AF109785.1 | - | 5 | 569 | |||
| AK036555.1 | - | 598 | ||||

Klrc2
Genomic Map