siRNA Details
| Gene Symbol: Krt27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010666.1 | NP_034796.1 | 5 | 901 | ||
| GenBank mRNA | AB013607.1 | - | 5 | 901 | ||
| AK077384.1 | - | 5 | 913 | |||
| BC140996.1 | - | 5 | 925 | 9055608 | ||
| BC140997.1 | - | 5 | 925 | 9055609 | ||

Krt27
Genomic Map