siRNA Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010581.3 | NP_034711.1 | 3 | 553 | ||
| GenBank mRNA | AB012693.1 | - | 3 | 492 | ||
| BC012667.1 | - | 3 | 487 | 4187965 | ||
| Z25524.1 | - | 3 | 553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd47| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010581.3 | NP_034711.1 | 3 | 553 | ||
| GenBank mRNA | AB012693.1 | - | 3 | 492 | ||
| BC012667.1 | - | 3 | 487 | 4187965 | ||
| Z25524.1 | - | 3 | 553 | |||