siRNA Details
| Gene Symbol: Kcnj5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010605.4 | NP_034735.3 | 3 | 937 | ||
| GenBank mRNA | AB019560.1 | - | 1 | 478 | ||
| AK137600.1 | - | 3 | 1051 | |||
| AK164058.1 | - | 3 | 938 | |||
| BC104330.1 | - | 1 | 496 | 40045462 | ||
| BC104331.1 | - | 1 | 496 | 40045465 | ||
| BC114976.1 | - | 1 | 493 | 40045466 | ||
| U33631.1 | - | 1 | 498 | |||

Kcnj5
Genomic Map