siRNA Details
| Gene Symbol: Klk1b24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010643.1 | NP_034773.1 | 525 | |||
| GenBank mRNA | AB039277.1 | - | 525 | |||
| AF019086.1 | - | 97 | ||||
| BC146015.1 | - | 4 | 552 | 40131318 | ||
| BC146019.1 | - | 4 | 552 | 40131322 | ||

Klk1b24
Genomic Map