siRNA Details
| Gene Symbol: Sspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010656.2 | NP_034786.1 | 2, 3 | 320 | ||
| GenBank mRNA | AK148179.1 | - | 2, 3 | 322 | ||
| AK162864.1 | - | 2, 3 | 345 | |||
| AK163041.1 | - | 2, 3 | 346 | |||
| AK164430.1 | - | 2, 3 | 346 | |||
| AK164529.1 | - | 2, 3 | 292 | |||
| BC021484.1 | - | 2, 3 | 345 | 5344514 | ||
| U02487.1 | - | 2, 3 | 320 | |||

Sspn
Genomic Map