siRNA Details
| Gene Symbol: Mafb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010658.3 | NP_034788.1 | 1 | 1282 | ||
| GenBank mRNA | AK131926.1 | - | 1 | 428 | ||
| AK132425.1 | - | 1 | 1283 | |||
| AK143209.1 | - | 1 | 1284 | |||
| AK154830.1 | - | 1 | 644 | |||
| BC016434.1 | - | 1 | 985 | 4485468 | ||
| BC038256.1 | - | 1 | 1230 | 5097650 | ||
| L36435.1 | - | 1 | 1062 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_019316, U56241.

Mafb
Genomic Map