siRNA Details
| Gene Symbol: Invs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010569.4 | NP_034699.3 | 3, 4 | 521 | ||
| GenBank mRNA | AF034860.3 | - | 3, 4 | 463 | ||
| AJ010902.1 | - | 3, 4 | 462 | |||
| AK041671.1 | - | 3, 4 | 456 | |||
| BX528321.1 | - | 3, 4 | 502 | |||

Invs
Genomic Map