siRNA Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010581.3 | NP_034711.1 | 10 | 1234 | ||
| GenBank mRNA | AB012693.1 | - | 10 | 1173 | ||
| AK151922.1 | - | 9 | 1117 | |||
| BC009094.1 | - | 8 | 800 | |||
| BC012667.1 | - | 10 | 1168 | 4187965 | ||
| Z25524.1 | - | 10 | 1234 | |||

Cd47
Genomic Map