siRNA Details
| Gene Symbol: Itih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010582.3 | NP_034712.2 | 7 | 784 | ||
| GenBank mRNA | AK133782.1 | - | 7 | 683 | ||
| AK149533.1 | - | 8 | 829 | |||
| BC034341.1 | - | 7 | 758 | 4211576 | ||
| X70392.1 | - | 7 | 770 | |||

Itih2
Genomic Map