siRNA Details
| Gene Symbol: Kap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010594.2 | NP_034724.1 | 4 | 459 | ||
| GenBank mRNA | AK142506.1 | - | 4 | 460 | ||
| AV000934.1 | - | 4 | 476 | |||
| BC013453.1 | - | 4 | 445 | 4221610 | ||
| M22810.1 | - | 4 | 458 | |||

Kap
Genomic Map