siRNA Details
| Gene Symbol: Kcnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159850.1 | NP_001153322.1 | 7 | 1659 | ||
| NM_001281847.1 | NP_001268776.1 | 7 | 1550 | |||
| NM_001281848.1 | NP_001268777.1 | 6 | 1391 | |||
| NM_010607.3 | NP_034737.2 | 7 | 1755 | |||
| GenBank mRNA | AK138427.1 | - | 7 | 1431 | ||
| AK140716.1 | - | 7 | 1430 | |||
| AK164197.1 | - | 6 | 1391 | |||
| BC062094.1 | - | 7 | 1373 | 6848897 | ||
| BQ572833.1 | - | 1 | 231 | |||
| U73488.2 | - | 8 | 1832 | |||

Kcnk2
Genomic Map