siRNA Details
| Gene Symbol: TAAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138327.2 | NP_612200.1 | 1 | 572 | ||
| GenBank mRNA | AY180374.1 | - | 1 | 549 | ||
| BC101825.1 | - | 1 | 572 | 8069331 | ||
| BC112194.1 | - | 1 | 584 | 8327662 | ||
More Information
Set Membership: |
|

TAAR1
Genomic Map