siRNA Details
| Gene Symbol: Klf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010638.4 | NP_034768.2 | 2 | 3239 | ||
| GenBank mRNA | AK028544.1 | - | 2 | 3242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf9| Gene Symbol: Klf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010638.4 | NP_034768.2 | 2 | 3239 | ||
| GenBank mRNA | AK028544.1 | - | 2 | 3242 | ||