siRNA Details
| Gene Symbol: Klra9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U49866.1 | - | 984 | |||
| U49867.1 | - | 984 | ||||
| Gene Symbol: Klra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC141217.1 | - | 914 | 9055829 | ||
| U10305.1 | - | 7 | 911 | |||
| U56405.1 | - | 7 | 870 | |||

Klra9
Genomic Map